missing translation for 'onlineSavingsMsg'
Learn More

Di(p-tolyl)phosphine Oxide 95.0+%, TCI America™

Catalog No. D45201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45201G 1 g
D45205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45201G Supplier TCI America Supplier No. D45201G

Chemical Identifiers

CAS 2409-61-2
Molecular Formula C14H14OP+
Molecular Weight (g/mol) 229.239
MDL Number MFCD01445489
InChI Key ZHIPXAFNKGZMSC-UHFFFAOYSA-N
Synonym bis p-tolyl phosphine oxide, di-p-tolylphosphine oxide, bis 4-methylphenyl phosphine oxide, di p-tolyl phosphine oxide, phosphine oxide, bis 4-methylphenyl, 1-methyl-4-4-methylphenylphosphoroso benzene, bis-p-tolylphosphine oxide, oxo bis-p-tolyl phosphonium, bis 4-tolyl phosphine oxide, ksc916e3n
PubChem CID 13357841
IUPAC Name bis(4-methylphenyl)-oxophosphanium
SMILES CC1=CC=C(C=C1)[P+](=O)C2=CC=C(C=C2)C

Specifications

Melting Point 96°C
Color White
Quantity 1 g
Formula Weight 230.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di(p-tolyl)phosphine Oxide