missing translation for 'onlineSavingsMsg'
Learn More

(-)-Di-p-toluoyl-L-tartaric Acid 98.0+%, TCI America™

Catalog No. p-7126704
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
D138725G 25 g
D1387250G 250 g
2 options

Catalog No. D138725G

Supplier: TCI America D138725G

Only null left

Chemical Identifiers

CAS 32634-66-5
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00064440
InChI Key CMIBUZBMZCBCAT-IYBDPMFKSA-N
Synonym 2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r
PubChem CID 12297760
IUPAC Name (2R,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 25 g
Formula Weight 386.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Di-p-toluoyl-L-tartaric Acid