missing translation for 'onlineSavingsMsg'
Learn More

(+)-Di-p-anisoyl-D-tartaric Acid 98.0+%, TCI America™

Catalog No. p-7127754
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34905G 5 g
D349025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34905G Supplier TCI America Supplier No. D34905G
Only null left

Chemical Identifiers

CAS 191605-10-4
Molecular Formula C20H18O10
Molecular Weight (g/mol) 418.35
MDL Number MFCD07368366
InChI Key KWWCVCFQHGKOMI-UHFFFAOYNA-N
Synonym (+)-Bis(4-methoxybenzoyl)-D-tartaric Acid
PubChem CID 9810979
IUPAC Name 2,3-bis(4-methoxybenzoyloxy)butanedioic acid
SMILES COC1=CC=C(C=C1)C(=O)OC(C(OC(=O)C1=CC=C(OC)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 176°C
Quantity 5 g
Formula Weight 418.35
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material (+)-Di-p-anisoyl-D-tartaric Acid