missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di(N-succinimidyl) Sebacate 98.0+%, TCI America™
Supplier: TCI America D40191G
Specifications
| Di(N-succinimidyl) Sebacate | |
| 169°C | |
| C18H24N2O8 | |
| 1 g | |
| XPKIVYVYXKPGBP-UHFFFAOYSA-N | |
| bis(2,5-dioxopyrrolidin-1-yl) decanedioate | |
| 3430836 | |
| ≥98.0% (HPLC,N) |
| 23024-29-5 | |
| White | |
| MFCD00216005 | |
| Sebacic Acid Di(N-succinimidyl) Ester, Di(N-succinimidyl) Decanedioate, Decanedioic Acid Di(N-succinimidyl) Ester | |
| O=C(CCCCCCCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O | |
| 396.40 | |
| 396.40 | |
| Crystalline Powder |
Chemical Identifiers
| 23024-29-5 | |
| 396.40 | |
| XPKIVYVYXKPGBP-UHFFFAOYSA-N | |
| 3430836 | |
| O=C(CCCCCCCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O |
| C18H24N2O8 | |
| MFCD00216005 | |
| Sebacic Acid Di(N-succinimidyl) Ester, Di(N-succinimidyl) Decanedioate, Decanedioic Acid Di(N-succinimidyl) Ester | |
| bis(2,5-dioxopyrrolidin-1-yl) decanedioate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer