missing translation for 'onlineSavingsMsg'
Learn More

Di(N-succinimidyl) Sebacate 98.0+%, TCI America™

Catalog No. D40195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40195G 5 g
D40191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40195G Supplier TCI America Supplier No. D40195G
Only null left

Chemical Identifiers

CAS 23024-29-5
Molecular Formula C18H24N2O8
Molecular Weight (g/mol) 396.40
MDL Number MFCD00216005
InChI Key XPKIVYVYXKPGBP-UHFFFAOYSA-N
Synonym Sebacic Acid Di(N-succinimidyl) Ester, Di(N-succinimidyl) Decanedioate, Decanedioic Acid Di(N-succinimidyl) Ester
PubChem CID 3430836
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) decanedioate
SMILES O=C(CCCCCCCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 396.40
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Di(N-succinimidyl) Sebacate
TSCA No
Recommended Storage Freezer