missing translation for 'onlineSavingsMsg'
Learn More

Di(N-succinimidyl) Carbonate 98.0+%, TCI America™

Catalog No. D16625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16625G 5 g
D166225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16625G Supplier TCI America Supplier No. D16625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74124-79-1
Molecular Formula C9H8N2O7
Molecular Weight (g/mol) 256.17
MDL Number MFCD00009767
InChI Key PFYXSUNOLOJMDX-UHFFFAOYSA-N
Synonym n,n'-disuccinimidyl carbonate, bis 2,5-dioxopyrrolidin-1-yl carbonate, n,n'-disuccinimidylcarbonate, di succinimido carbonate, disuccinimidyl carbonate, di n-succinimidyl carbonate, n,n-disuccinimidyl carbonate, dsc, disuccinimido carbonate, pubchem12700
PubChem CID 676246
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 215°C
Quantity 5 g
Formula Weight 256.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di(N-succinimidyl) Carbonate
TSCA No
Recommended Storage Refrigerator