missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl succinate, 98%

Catalog No. AAB2044109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2044109 10 g
AAB2044118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2044109 Supplier Thermo Scientific Chemicals Supplier No. B2044109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 925-15-5 | C10H18O4 | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 925-15-5
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00015213
InChI Key SZHZCPHKDJWHNG-UHFFFAOYSA-N
Synonym dipropyl succinate, di-n-propyl succinate, succinic acid, dipropyl ester, butanedioic acid, dipropyl ester, dipropylester kyseliny jantarove, dipropylester kyseliny jantarove czech, dipropyl butane-1,4-dioate, n-propyl succinate, 1,4-dipropyl butanedioate, succinic acid di-n-propyl ester
PubChem CID 13549
IUPAC Name dipropyl butanedioate
SMILES CCCOC(=O)CCC(=O)OCCC

Specifications

Density 1
Boiling Point 250°C to 252°C
Odor Pleasant
Quantity 10 g
Formula Weight 202.25
Percent Purity 98%
Chemical Name or Material Di-n-propyl succinate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-114-2
RTECSNumber WM7750000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only