Learn More
Di-n-propyl ether, 99%
CAS: 111-43-3 | C6H14O | 102.18 g/mol
$194.18 - $563.14
Chemical Identifiers
| CAS | 111-43-3 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.18 |
| MDL Number | MFCD00009376 |
| InChI Key | POLCUAVZOMRGSN-UHFFFAOYSA-N |
| Synonym | dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g |
| PubChem CID | 8114 |
| IUPAC Name | 1-propoxypropane |
| SMILES | CCCOCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2142314
|
Thermo Scientific Chemicals
B2142314 |
25 g |
Each for $194.18
|
|
|||||
|
AAB2142322
|
Thermo Scientific Chemicals
B2142322 |
100 g |
Each for $563.14
|
|
|||||
Description
Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.
Solubility
It is slightly soluble in water 3 g/L (20°C), soluble in alcohol and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids, halogens.
Chemical Identifiers
| 111-43-3 | |
| 102.18 | |
| POLCUAVZOMRGSN-UHFFFAOYSA-N | |
| 8114 | |
| CCCOCCC |
| C6H14O | |
| MFCD00009376 | |
| dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g | |
| 1-propoxypropane |
Specifications
| 111-43-3 | |
| 0.747 | |
| −28°C (−18°F) | |
| MFCD00009376 | |
| UN2384 | |
| It is slightly soluble in water 3g/L (20°C),soluble in alcohol and ether. | |
| CCCOCCC | |
| 102.18 | |
| 102.18 | |
| Di-n-propyl ether |
| -122°C | |
| 90°C to 91°C | |
| C6H14O | |
| 25 g | |
| dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g | |
| POLCUAVZOMRGSN-UHFFFAOYSA-N | |
| 1-propoxypropane | |
| 8114 | |
| 99% |
Safety and Handling
GHS H Statement
H225-H336
Highly flammable liquid and vapour.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P304+P340-P312-P370+P378q-P372-P374-P380-P501c
H225-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DI-N-PROPYL ETHER
EINECSNumber : 203-869-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only