missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl ether, 99%

Catalog No. AAB2142314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2142314 25 g
AAB2142322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2142314 Supplier Thermo Scientific Chemicals Supplier No. B2142314

CAS: 111-43-3 | C6H14O | 102.18 g/mol

Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.

Solubility
It is slightly soluble in water 3 g/L (20°C), soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids, halogens.

Chemical Identifiers

CAS 111-43-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00009376
InChI Key POLCUAVZOMRGSN-UHFFFAOYSA-N
Synonym dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g
PubChem CID 8114
IUPAC Name 1-propoxypropane
SMILES CCCOCCC

Specifications

Melting Point -122°C
Density 0.747
Boiling Point 90°C to 91°C
Flash Point −28°C (−18°F)
Quantity 25 g
UN Number UN2384
Solubility Information It is slightly soluble in water 3g/L (20°C),soluble in alcohol and ether.
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material Di-n-propyl ether
Hazard Category H225-H336
Hazard Statement GHS H Statement
H225-H336
Highly flammable liquid and vapour.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P304+P340-P312-P370+P378q-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DI-N-PROPYL ETHER
EINECSNumber 203-869-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only