missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyltin oxide

Catalog No. p-7080334
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA7119314 25 g
AA7119322 100 g
AA7119336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA7119314 Supplier Thermo Scientific Chemicals Supplier No. 07119314

CAS: 870-08-6 | C16H34OSn | 361.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 870-08-6
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00013839
InChI Key LQRUPWUPINJLMU-UHFFFAOYSA-N
Synonym di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin
PubChem CID 93563
IUPAC Name dioctyl(oxo)tin
SMILES CCCCCCCC[Sn](=O)CCCCCCCC

Specifications

Melting Point 245°C to 248°C (decomposition)
Color White
Quantity 25 g
Formula Weight 361.14
Physical Form Powder
Chemical Name or Material Di-n-octyltin oxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-791-1
RTECSNumber WH7620000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only