missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-octyltin Oxide 98.0+%, TCI America™
Supplier: TCI America D1373500G
Specifications
| Di-n-octyltin Oxide | |
| White | |
| MFCD00013839 | |
| di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin | |
| CCCCCCCC[Sn](=O)CCCCCCCC | |
| 361.16 | |
| 361.16 | |
| Crystalline Powder |
| 870-08-6 | |
| C16H34OSn | |
| 500 g | |
| LQRUPWUPINJLMU-UHFFFAOYSA-N | |
| dioctylstannanone | |
| 93563 | |
| ≥98.0% (W) |
Chemical Identifiers
| 870-08-6 | |
| 361.16 | |
| LQRUPWUPINJLMU-UHFFFAOYSA-N | |
| 93563 | |
| CCCCCCCC[Sn](=O)CCCCCCCC |
| C16H34OSn | |
| MFCD00013839 | |
| di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin | |
| dioctylstannanone |
Safety and Handling
RTECSNumber : WH7620000
TSCA : Yes