missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyltin Oxide 98.0+%, TCI America™

Catalog No. D137325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D137325G 25 g
D1373500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D137325G Supplier TCI America Supplier No. D137325G

Chemical Identifiers

CAS 870-08-6
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00013839
InChI Key LQRUPWUPINJLMU-UHFFFAOYSA-N
Synonym di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin
PubChem CID 93563
IUPAC Name dioctylstannanone
SMILES CCCCCCCC[Sn](=O)CCCCCCCC

Specifications

Color White
Quantity 25 g
Formula Weight 361.16
Percent Purity ≥98.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Di-n-octyltin Oxide
RTECSNumber WH7620000
TSCA Yes