missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octylamine 96.0+%, TCI America™

Catalog No. D027425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D027425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D027425ML Supplier TCI America Supplier No. D027425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1120-48-5
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.463
MDL Number MFCD00009557
InChI Key LAWOZCWGWDVVSG-UHFFFAOYSA-N
Synonym dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine
PubChem CID 3094
IUPAC Name N-octyloctan-1-amine
SMILES CCCCCCCCNCCCCCCCC

Specifications

Melting Point 15°C
Color Yellow
Boiling Point 298°C
Quantity 25 mL
UN Number 2735
Formula Weight 241.46
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Di-n-octylamine
EINECSNumber (2)-0176&(2)-0138
TSCA Yes