missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octylamine, 95%

Catalog No. p-3736358
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117080250 25 g Glass bottle
AC117081000 100 g Glass bottle
AC117085000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC117080250 Supplier Thermo Scientific Chemicals Supplier No. 117080250
Only null left

CAS: 1120-48-5 | C16H35N | 241.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1120-48-5
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
MDL Number MFCD00009557
InChI Key LAWOZCWGWDVVSG-UHFFFAOYSA-N
Synonym dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine
PubChem CID 3094
IUPAC Name N-octyloctan-1-amine
SMILES CCCCCCCCNCCCCCCCC

Specifications

Melting Point 14°C to 15°C
Color Colorless to Yellow
Density 0.7900g/mL
Boiling Point 297°C to 298°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4412 to 1.4432
Linear Formula [CH3(CH2)7]2NH
Quantity 25 g
Beilstein 04, 196
Specific Gravity 0.79
Formula Weight 241.46
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Di-n-octylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 214-311-6

RUO – Research Use Only