missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octylamine, 96%

Catalog No. AAA1801722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1801722 100 g
AAA1801714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1801722 Supplier Thermo Scientific Chemicals Supplier No. A1801722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1120-48-5 | C16H35N | 241.463 g/mol

Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.

Solubility
Immiscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.

Chemical Identifiers

CAS 1120-48-5
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.463
MDL Number MFCD00009557
InChI Key LAWOZCWGWDVVSG-UHFFFAOYSA-N
Synonym dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine
PubChem CID 3094
IUPAC Name N-octyloctan-1-amine
SMILES CCCCCCCCNCCCCCCCC

Specifications

Melting Point 13°C to 15°C
Density 0.799
Boiling Point 297°C to 298°C
Flash Point 142°C (287°F)
Odor Amine-like
Refractive Index 1.4432
Linear Formula [CH3(CH2)7]2NH
Quantity 100 g
UN Number UN2735
Beilstein 1748376
Solubility Information Immiscible in water.
Formula Weight 241.46
Percent Purity 98%
Chemical Name or Material Di-n-octylamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 214-311-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only