missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl Sulfone 98.0+%, TCI America™

Catalog No. D31615G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31615G Supplier TCI America Supplier No. D31615G
Only null left

Chemical Identifiers

CAS 7726-20-7
Molecular Formula C16H34O2S
Molecular Weight (g/mol) 290.51
MDL Number MFCD00039760
InChI Key TZPCGEVFZZYCIY-UHFFFAOYSA-N
Synonym n-Octyl Sulfone
PubChem CID 3798558
IUPAC Name 1-(octane-1-sulfonyl)octane
SMILES CCCCCCCCS(=O)(=O)CCCCCCCC

Specifications

Melting Point 78°C
Color White
Quantity 5 g
Formula Weight 290.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di-n-octyl Sulfone