missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexylamine, 98+%

Catalog No. AAL06326AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL06326AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL06326AE Supplier Thermo Scientific Chemicals Supplier No. L06326AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 143-16-8 | C12H27N | 185.355 g/mol

Di-n-hexylamine is used in organic synthesis and involved in the preparation of succinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-hexylamine is used in organic synthesis and involved in the preparation of succinic acid.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 143-16-8
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.355
MDL Number MFCD00009521
InChI Key PXSXRABJBXYMFT-UHFFFAOYSA-N
Synonym dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
PubChem CID 8920
IUPAC Name N-hexylhexan-1-amine
SMILES CCCCCCNCCCCCC

Specifications

Melting Point 3°C
Density 0.79
Boiling Point 236°C
Flash Point 104°C (219°F)
Refractive Index 1.4335
Quantity 100 mL
UN Number UN2927
Beilstein 1738519
Solubility Information Miscible with water.
Formula Weight 185.36
Percent Purity ≥98%
Chemical Name or Material Di-n-hexylamine
Show More Show Less
Hazard Category H301-H310+H330-H314
Hazard Statement GHS H Statement
H310-H330-H314-H318-H302
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P261-P262-P264b-P270-P271-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 205-588-4
RTECSNumber IH6600000
TSCA Yes

RUO – Research Use Only