Learn More
Di-n-decyl dimethylammonium bromide, 80% aq. gel
CAS: 2390-68-3 | C22H48BrN | 406.537 g/mol
$118.99 - $421.90
Chemical Identifiers
| CAS | 2390-68-3 |
|---|---|
| Molecular Formula | C22H48BrN |
| Molecular Weight (g/mol) | 406.537 |
| MDL Number | MFCD00012194 |
| InChI Key | UMGXUWVIJIQANV-UHFFFAOYSA-M |
| Synonym | didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide |
| PubChem CID | 16957 |
| IUPAC Name | didecyl(dimethyl)azanium;bromide |
| SMILES | CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2483222
|
Thermo Scientific Chemicals
B2483222 |
100 g |
Each for $118.99
|
|
|||||
|
AAB2483236
|
Thermo Scientific Chemicals
B2483236 |
500 g |
Each for $421.90
|
|
|||||
Description
Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.
Solubility
Soluble in methanol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 2390-68-3 | |
| 406.537 | |
| UMGXUWVIJIQANV-UHFFFAOYSA-M | |
| 16957 | |
| CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-] |
| C22H48BrN | |
| MFCD00012194 | |
| didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide | |
| didecyl(dimethyl)azanium;bromide |
Specifications
| 2390-68-3 | |
| [CH3(CH2)9]2N(CH3)2Br | |
| MFCD00012194 | |
| didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide | |
| UMGXUWVIJIQANV-UHFFFAOYSA-M | |
| didecyl(dimethyl)azanium;bromide | |
| 16957 | |
| Di-n-decyl dimethylammonium bromide, 80% aq. gel |
| C22H48BrN | |
| 1.457 | |
| 100 g | |
| Soluble in methanol. | |
| CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-] | |
| 406.537 | |
| 406.54 |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 219-234-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only