missing translation for 'onlineSavingsMsg'
Learn More

Di-n-decyl dimethylammonium bromide, 80% aq. gel

Catalog No. AAB2483222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2483222 100 g
AAB2483236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2483222 Supplier Thermo Scientific Chemicals Supplier No. B2483222

CAS: 2390-68-3 | C22H48BrN | 406.537 g/mol

Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 2390-68-3
Molecular Formula C22H48BrN
Molecular Weight (g/mol) 406.537
MDL Number MFCD00012194
InChI Key UMGXUWVIJIQANV-UHFFFAOYSA-M
Synonym didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide
PubChem CID 16957
IUPAC Name didecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-]

Specifications

Refractive Index 1.457
Linear Formula [CH3(CH2)9]2N(CH3)2Br
Quantity 100 g
Solubility Information Soluble in methanol.
Formula Weight 406.54
Chemical Name or Material Di-n-decyl dimethylammonium bromide, 80% aq. gel
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 219-234-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only