missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin oxide

Catalog No. p-7049354
Change view
Click to view available options
Quantity:
50 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1411718 50 g
AA14117A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1411718 Supplier Thermo Scientific Chemicals Supplier No. 01411718
Only null left

CAS: 818-08-6 | C8H18OSn | 248.941 g/mol

Di-n-butyltin oxide is used as a reagent and a catalyst. It is particularly useful in regioselective alkylation, acylation and sulfonation reactions for starting materials containing alcohol functional groups. It is used in studies pertaining to discovering novel antitumor agents, antifungal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 818-08-6
Molecular Formula C8H18OSn
Molecular Weight (g/mol) 248.941
MDL Number MFCD00001992
InChI Key JGFBRKRYDCGYKD-UHFFFAOYSA-N
Synonym dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
PubChem CID 61221
IUPAC Name dibutyl(oxo)tin
SMILES CCCC[Sn](=O)CCCC

Specifications

Melting Point >300°C
Color White
Density 1.58 g/mL
Odor Odorless
Linear Formula [CH3(CH2)3]2SnO
Quantity 50 g
UN Number UN3146
Beilstein 4126243
Solubility Information Soluble in water 4.0g/L @ 20°C.
Formula Weight 248.92
Physical Form Powder
Chemical Name or Material Di-n-butyltin oxide
Show More Show Less
Hazard Category H301-H315-H317-H318-H341-H360-H370-H372
Hazard Statement GHS H Statement
H300-H319
Fatal if swallowed.
Causes serious eye irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P307+P311-P310-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 212-449-1
RTECSNumber WH7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only