Learn More
Di-n-butyltin oxide, 98%
CAS: 818-08-6 | C8H18OSn | 248.941 g/mol
Supplier: Thermo Scientific Chemicals L1449136
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Di-n-butyltin oxide | |
| >300°C | |
| Odorless | |
| [CH3(CH2)3]2SnO | |
| 500 g | |
| 4126243 | |
| JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
| dibutyl(oxo)tin | |
| 61221 | |
| 98% |
| 818-08-6 | |
| 1.58 | |
| C8H18OSn | |
| MFCD00001992 | |
| UN3146 | |
| dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
| CCCC[Sn](=O)CCCC | |
| 248.941 | |
| 248.94 |
Chemical Identifiers
| 818-08-6 | |
| 248.941 | |
| JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
| 61221 | |
| CCCC[Sn](=O)CCCC |
| C8H18OSn | |
| MFCD00001992 | |
| dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
| dibutyl(oxo)tin |
Safety and Handling
GHS H Statement
H300-H319
Fatal if swallowed.
Causes serious eye irritation.
P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P307+P311-P310-P330-P333+P313-P362-P501c
H301-H315-H317-H318-H341-H360-H370-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber : 212-449-1
RTECSNumber : WH7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only