missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin oxide, 98%

Catalog No. AAL1449122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1449122 100 g
AAL1449136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL1449122 Supplier Thermo Scientific Chemicals Supplier No. L1449122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 818-08-6 | C8H18OSn | 248.941 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 818-08-6
Molecular Formula C8H18OSn
Molecular Weight (g/mol) 248.941
MDL Number MFCD00001992
InChI Key JGFBRKRYDCGYKD-UHFFFAOYSA-N
Synonym dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
PubChem CID 61221
IUPAC Name dibutyl(oxo)tin
SMILES CCCC[Sn](=O)CCCC

Specifications

Melting Point >300°C
Density 1.58
Odor Odorless
Linear Formula [CH3(CH2)3]2SnO
Quantity 100 g
UN Number UN3146
Beilstein 4126243
Formula Weight 248.94
Percent Purity 98%
Chemical Name or Material Di-n-butyltin oxide
Hazard Category H301-H315-H317-H318-H341-H360-H370-H372
Hazard Statement GHS H Statement
H300-H319
Fatal if swallowed.
Causes serious eye irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P307+P311-P310-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 212-449-1
RTECSNumber WH7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only