Learn More
Di-n-butylmagnesium, 0.5M solution in heptane, AcroSeal™
CAS: 1191-47-5 | C8H18Mg | 138.54 g/mol
$305.24 - $305.24
Chemical Identifiers
| CAS | 1191-47-5 |
|---|---|
| Molecular Formula | C8H18Mg |
| Molecular Weight (g/mol) | 138.54 |
| MDL Number | MFCD00015225 |
| InChI Key | KJJBSBKRXUVBMX-UHFFFAOYSA-N |
| Synonym | dibutylmagnesium, magnesium, dibutyl, di-n-butylmagnesium, magnesium,butyl 1-methylpropyl, dibutyl magnesium, di-n-butyl magnesium, di-n-butylmagnesium solution, odhfjiddbsdwnu-uhfffaoysa-n, di-n-butylmagnesium, 0,5m in heptane, di-n-butylmagnesium solution, 1.0 m in heptane |
| PubChem CID | 70929 |
| IUPAC Name | magnesium;butane |
| SMILES | [Mg++].CCC[CH2-].CCC[CH2-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC377771000
|
Thermo Scientific Chemicals
377771000 |
100 mL | AcroSeal™ Glass bottle |
Each for $305.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1191-47-5 | |
| 138.54 | |
| KJJBSBKRXUVBMX-UHFFFAOYSA-N | |
| 70929 | |
| [Mg++].CCC[CH2-].CCC[CH2-] |
| C8H18Mg | |
| MFCD00015225 | |
| dibutylmagnesium, magnesium, dibutyl, di-n-butylmagnesium, magnesium,butyl 1-methylpropyl, dibutyl magnesium, di-n-butyl magnesium, di-n-butylmagnesium solution, odhfjiddbsdwnu-uhfffaoysa-n, di-n-butylmagnesium, 0,5m in heptane, di-n-butylmagnesium solution, 1.0 m in heptane | |
| magnesium;butane |
Specifications
| 1191-47-5,142-82-5,97-93-8 | |
| 0.7300g/mL | |
| AcroSeal™ Glass bottle | |
| (C4H9)2Mg | |
| 100 mL | |
| dibutylmagnesium, magnesium, dibutyl, di-n-butylmagnesium, magnesium,butyl 1-methylpropyl, dibutyl magnesium, di-n-butyl magnesium, di-n-butylmagnesium solution, odhfjiddbsdwnu-uhfffaoysa-n, di-n-butylmagnesium, 0,5m in heptane, di-n-butylmagnesium solution, 1.0 m in heptane | |
| KJJBSBKRXUVBMX-UHFFFAOYSA-N | |
| magnesium;butane | |
| 70929 | |
| 138.54 |
| -71.0°C | |
| −4°C | |
| C8H18Mg | |
| MFCD00015225 | |
| 0.73 | |
| Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons | |
| [Mg++].CCC[CH2-].CCC[CH2-] | |
| 138.54 | |
| 0.9 mPa.s (22°C) | |
| Di-n-butylmagnesium |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
In contact with water releases
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 214-736-7
RUO – Research Use Only