missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylamine, for HPLC

Catalog No. p-3586424
Change view
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC390610100 10 mL Glass bottle
AC390611000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC390610100 Supplier Thermo Scientific Chemicals Supplier No. 390610100
Only null left

CAS: 111-92-2 | C8H19N | 129.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-92-2
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.24
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

Melting Point -62.0°C
Color White
Density 0.7600g/mL
Boiling Point 159.0°C
Flash Point 39°C
Packaging Glass bottle
Refractive Index 1.4160 to 1.4180
Linear Formula [CH3(CH2)3]2NH
Quantity 10 mL
Residue after Evaporation 0.0005% max.
Specific Gravity 0.76
Merck Index 15, 3037
Solubility Information Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons
Transmittance 99 (c=0.01M water, 254nm) min.
Viscosity 0.9 mPa.s (20°C)
Formula Weight 129.24
Percent Purity ≥99% (GC)
Grade HPLC
Physical Form Crystalline Free Flowing Powder
Chemical Name or Material Di-n-butylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre
EINECSNumber 203-921-8

RUO – Research Use Only