missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl sulfate, 95%

Catalog No. p-7046016
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1281309 10 g
AAL1281318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1281309 Supplier Thermo Scientific Chemicals Supplier No. L1281309
Only null left

CAS: 625-22-9 | C8H18O4S | 210.288 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-22-9
Molecular Formula C8H18O4S
Molecular Weight (g/mol) 210.288
MDL Number MFCD00059420
InChI Key LMEDOLJKVASKTP-UHFFFAOYSA-N
Synonym dibutylsulfate, di-n-butylsulfat, sulfuric acid, dibutyl ester, di-n-butylsulfate, di-n-butyl sulfate, dibutyl sulphate, di-n-butylsulfat german, sulfuric acid dibutyl ester, sulphuric acid dibutyl ester, dibutylsulfat
PubChem CID 12239
IUPAC Name dibutyl sulfate
SMILES CCCCOS(=O)(=O)OCCCC

Specifications

Density 1.06
Boiling Point 115°C to 116°C (6 mmHg)
Refractive Index 1.4205
Linear Formula (CH3CH2CH2CH2O)2SO2
Quantity 10 g
Beilstein 1707498
Formula Weight 210.29
Percent Purity 95%
Chemical Name or Material Di-n-butyl sulfate
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber WS7700000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only