Learn More
Di-n-butyl sebacate, 90+%
CAS: 109-43-3 | C18H34O4 | 314.466 g/mol
Supplier: Thermo Scientific Chemicals A1823136
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Di-n-butyl sebacate | |
| -11°C | |
| 344°C to 345°C | |
| C18H34O4 | |
| MFCD00027218 | |
| 1798308 | |
| PYGXAGIECVVIOZ-UHFFFAOYSA-N | |
| dibutyl decanedioate | |
| 7986 | |
| 94% |
| 109-43-3 | |
| 0.94 | |
| 178°C (352°F) | |
| 1.441 | |
| 500 g | |
| dibutyl sebacate, butyl sebacate, di-n-butyl sebacate, decanedioic acid, dibutyl ester, polycizer dbs, kodaflex dbs, dibutyl sebacinate, staflex dbs, sebacic acid, dibutyl ester, monoplex dbs | |
| CCCCOC(=O)CCCCCCCCC(=O)OCCCC | |
| 314.466 | |
| 314.47 |
Chemical Identifiers
| 109-43-3 | |
| 314.466 | |
| PYGXAGIECVVIOZ-UHFFFAOYSA-N | |
| 7986 | |
| CCCCOC(=O)CCCCCCCCC(=O)OCCCC |
| C18H34O4 | |
| MFCD00027218 | |
| dibutyl sebacate, butyl sebacate, di-n-butyl sebacate, decanedioic acid, dibutyl ester, polycizer dbs, kodaflex dbs, dibutyl sebacinate, staflex dbs, sebacic acid, dibutyl ester, monoplex dbs | |
| dibutyl decanedioate |
Safety and Handling
EINECSNumber : 203-672-5
RTECSNumber : VS1150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only