missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl sebacate, 90+%

Catalog No. AAA1823122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1823122 100 g
AAA1823136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1823122 Supplier Thermo Scientific Chemicals Supplier No. A1823122

CAS: 109-43-3 | C18H34O4 | 314.466 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-43-3
Molecular Formula C18H34O4
Molecular Weight (g/mol) 314.466
MDL Number MFCD00027218
InChI Key PYGXAGIECVVIOZ-UHFFFAOYSA-N
Synonym dibutyl sebacate, butyl sebacate, di-n-butyl sebacate, decanedioic acid, dibutyl ester, polycizer dbs, kodaflex dbs, dibutyl sebacinate, staflex dbs, sebacic acid, dibutyl ester, monoplex dbs
PubChem CID 7986
IUPAC Name dibutyl decanedioate
SMILES CCCCOC(=O)CCCCCCCCC(=O)OCCCC

Specifications

Melting Point -11°C
Density 0.94
Boiling Point 344°C to 345°C
Flash Point 178°C (352°F)
Refractive Index 1.441
Quantity 100 g
Beilstein 1798308
Formula Weight 314.47
Percent Purity 94%
Chemical Name or Material Di-n-butyl sebacate
EINECSNumber 203-672-5
RTECSNumber VS1150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only