missing translation for 'onlineSavingsMsg'
Learn More

Di-mu-chlorobis[5-hydroxy-2-[1-(hydroxyimino)ethyl]phenyl]palladium(II) Dimer 95.0+%, TCI America™

Catalog No. p-7127929
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3807250MG 250 mg
D38071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3807250MG Supplier TCI America Supplier No. D3807250MG
Only null left

Chemical Identifiers

CAS 419581-64-9
Molecular Formula C16H16Cl2N2O4Pd2
Molecular Weight (g/mol) 584.054
MDL Number MFCD08705289
InChI Key OQQFWDBYVLTJST-ALDPEWTMSA-L
Synonym Najera Catalyst II
PubChem CID 134128703
IUPAC Name chloropalladium(1+);4-[1-(hydroxyamino)ethylidene]cyclohexa-2,5-dien-1-one;4-[(Z)-N-hydroxy-C-methylcarbonimidoyl]benzene-5-id-1-ol
SMILES CC(=C1C=CC(=O)C=[C-]1)NO.CC(=NO)C1=CC=C(C=[C-]1)O.Cl[Pd+].Cl[Pd+]

Specifications

Melting Point 255°C
Color Yellow
Quantity 250 mg
Formula Weight 584.05
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-mu-chlorobis[5-hydroxy-2-[1-(hydroxyimino)ethyl]phenyl]palladium(II) Dimer