missing translation for 'onlineSavingsMsg'
Learn More

Di-mu-chlorobis[2-[(dimethylamino)methyl]-4,6-dimethoxyphenyl-C,N]dipalladium(II) 98.0+%, TCI America™

Catalog No. D41121G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41121G Supplier TCI America Supplier No. D41121G
Only null left

Chemical Identifiers

CAS 18987-71-8
Molecular Formula C22H38Cl2N2O4Pd2+2
Molecular Weight (g/mol) 678.296
InChI Key ZTGHLWXKBFAGLC-UHFFFAOYSA-N
PubChem CID 131674810
IUPAC Name dichloranium;1-(3,5-dimethoxyphenyl)-N,N-dimethylmethanamine;palladium
SMILES CN(C)CC1=CC(=CC(=C1)OC)OC.CN(C)CC1=CC(=CC(=C1)OC)OC.[ClH2+].[ClH2+].[Pd].[Pd]

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 672.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-mu-chlorobis[2-[(dimethylamino)methyl]-4,6-dimethoxyphenyl-C,N]dipalladium(II)