missing translation for 'onlineSavingsMsg'
Learn More

Di-^m-chlorobis(norbornadiene)dirhodium(I), Rh 44% min

CAS: 12257-42-0 | C14H16Cl2Rh2 | 460.99 g/mol

$1078.60 - $1078.60

Chemical Identifiers

CAS 12257-42-0
Molecular Formula C14H16Cl2Rh2
Molecular Weight (g/mol) 460.99
MDL Number MFCD00198060
InChI Key RXDWVIOULPVOEO-UHFFFAOYSA-L
Synonym bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer, bicyclo 2.2.1 hepta-2,5-diene; rhodium 2+ ; dichloride, bis 2,3,5,6-eta-bicyclo 2.2.1 hepta-2,5-diene di-mu-chlorodirhodium
PubChem CID 114600
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1C2C=CC1C=C2.C1C2C=CC1C=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1046702
View Documents
Thermo Scientific Chemicals
01046702
0.5 g
Each for $1,078.60
Only null left
 
Description

Description

Di-μ-chlorobis(norbornadiene)dirhodium(I) is used as catalysts in olefin hydroformylation, olefin hydrogenation and olefin isomerization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12257-42-0
460.99
RXDWVIOULPVOEO-UHFFFAOYSA-L
114600
C14H16Cl2Rh2
MFCD00198060
bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer, bicyclo 2.2.1 hepta-2,5-diene; rhodium 2+ ; dichloride, bis 2,3,5,6-eta-bicyclo 2.2.1 hepta-2,5-diene di-mu-chlorodirhodium
[Cl-].[Cl-].[Rh+].[Rh+].C1C2C=CC1C=C2.C1C2C=CC1C=C2

Specifications

12257-42-0
C14H16Cl2Rh2
0.5 g
Slightly soluble in acetone,chloroform,methanol and insoluble in water.
[Cl-].[Cl-].[Rh+].[Rh+].C1C2C=CC1C=C2.C1C2C=CC1C=C2
460.99
461.01
Powder
Odorless
MFCD00198060
bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer, bicyclo 2.2.1 hepta-2,5-diene; rhodium 2+ ; dichloride, bis 2,3,5,6-eta-bicyclo 2.2.1 hepta-2,5-diene di-mu-chlorodirhodium
RXDWVIOULPVOEO-UHFFFAOYSA-L
bicyclo[2.2.1]hepta-2,5-diene;rhodium(2+);dichloride
114600
Rh 44% min.
Di-mu-chlorobis(norbornadiene)dirhodium(I)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 235-510-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only