missing translation for 'onlineSavingsMsg'
Learn More

di-Iodine Pentoxide, Granular (0.5 to 2.5mm), For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1003580100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1003580100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1003580100 Supplier Technidata Supplier No. 1.00358.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12029-98-0
Molecular Formula I2O5
Molecular Weight (g/mol) 333.804
InChI Key BIZCJSDBWZTASZ-UHFFFAOYSA-N
Synonym iodine pentoxide, iodopentoxide, iodic anhydride, iodine v oxide, diiodine pentaoxide, unii-upu35c0q0n, upu35c0q0n, iodine oxide, jodpentoxyd, iodine pentaoxide
PubChem CID 159402
ChEBI CHEBI:29914
IUPAC Name iodyl iodate
SMILES O=I(=O)OI(=O)=O

Specifications

Physical Form Solid
Quantity 100 g
UN Number UN 1479
Formula Weight 333.81
Odor Odorless
Packaging Glass bottle
Appearance Light yellow solid
Chemical Name or Material di-Iodine Pentoxide
Hazard Category H272: May intensify fire; oxidiser.
H315: Causes skin irritation.
H319: Causes serious eye irritation.