missing translation for 'onlineSavingsMsg'
Learn More

Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%

CAS: 30698-64-7 | C12H36Cl2Cu2N4O2 | 466.44 g/mol

$474.34 - $474.34

Chemical Identifiers

CAS 30698-64-7
Molecular Formula C12H36Cl2Cu2N4O2
Molecular Weight (g/mol) 466.44
MDL Number MFCD01321163
InChI Key VVXKYYDFGPZSOZ-UHFFFAOYSA-L
Synonym cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
PubChem CID 11190467
IUPAC Name chlorocopper;N,N,N',N'-tetramethylethane-1,2-diamine;dihydrate
SMILES O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC348980250
SDS View Documents
Thermo Scientific Chemicals
348980250
25 g Glass bottle
Each for $474.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

30698-64-7
466.44
VVXKYYDFGPZSOZ-UHFFFAOYSA-L
11190467
O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
C12H36Cl2Cu2N4O2
MFCD01321163
cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
chlorocopper;N,N,N',N'-tetramethylethane-1,2-diamine;dihydrate

Specifications

30698-64-7
Purple
97.5% min. (Argentometry)
C12H36Cl2Cu2N4O2
25 g
Solubility in water: insoluble in water
O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
466.44
464.42
Powder
136.0°C to 138.0°C
Authentic
Glass bottle
MFCD01321163
cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
VVXKYYDFGPZSOZ-UHFFFAOYSA-L
chlorocopper;N,N,N',N'-tetramethylethane-1,2-diamine;dihydrate
11190467
98%
Di-μ-hydroxo-bis-[(N,N,N′,N′-tetramethylethylenediamine)copper(II)]chloride
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only