missing translation for 'onlineSavingsMsg'
Learn More

di-Ammonium Oxalate Monohydrate, MilliporeSigma™

Catalog No. M1011921000
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1011921000 1 kg Plastic Bottle
M1011920250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1011921000 Supplier Technidata Supplier No. 1.01192.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Physical Form Solid
Quantity 1 kg
Solubility Information Soluble in water: 45g/L at 20°C
Formula Weight 142.11
Percent Purity 99.5% to 101.0%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Density 1.50g/cm3 at 20°C
Chemical Name or Material di-Ammonium oxalate monohydrate
Hazard Category H302: Harmful if swallowed
Hazard Statement H312: Harmful in contact with skin
EINECSNumber 214-202-3