missing translation for 'onlineSavingsMsg'
Learn More
Learn More
di-Ammonium Hydrogen Phosphate, MilliporeSigma™
For analysis EMSURE™ ACS, Reag. Ph Eur
$342.73 - $342.73
Chemical Identifiers
| CAS | 7783-28-0 |
|---|---|
| Molecular Formula | H9N2O4P |
| Molecular Weight (g/mol) | 132.06 |
| MDL Number | MFCD00010891 |
| InChI Key | MNNHAPBLZZVQHP-UHFFFAOYSA-N |
| Synonym | diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate |
| PubChem CID | 24540 |
| ChEBI | CHEBI:63051 |
| IUPAC Name | phosphoric acid diamine |
| SMILES | N.N.OP(O)(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1012070500
|
Technidata
1012070500 |
500 g | Plastic Bottle |
Each for $342.73
|
|
||||
Chemical Identifiers
| 7783-28-0 | |
| 132.06 | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| 24540 | |
| phosphoric acid diamine |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| CHEBI:63051 | |
| N.N.OP(O)(O)=O |
Specifications
| 7783-28-0 | |
| H9N2O4P | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| phosphoric acid diamine | |
| 24540 | |
| 132.05 | |
| Plastic Bottle | |
| di-Ammonium hydrogen phosphate |
| 500 g | |
| MFCD00010891 | |
| Soluble in water: 690g/L at 20°C | |
| N.N.OP(O)(O)=O | |
| 132.06 | |
| CHEBI:63051 | |
| Stinging | |
| 1.619g/cm3 at 20°C |
Safety and Handling
EINECSNumber : 231-987-8
Recommended Storage : Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry