missing translation for 'onlineSavingsMsg'
Learn More

Di-2-pyridyl ketone, 98%

Catalog No. AC155670050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155670050 5 g Glass bottle
AC155670250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155670050 Supplier Thermo Scientific Chemicals Supplier No. 155670050

CAS: 19437-26-4 | C11H8N2O | 184.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19437-26-4
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.2
MDL Number MFCD00006288
InChI Key QPOWUYJWCJRLEE-UHFFFAOYSA-N
Synonym di-2-pyridyl ketone, di pyridin-2-yl methanone, bis 2-pyridyl ketone, 2,2'-carbonyldipyridine, di 2-pyridyl ketone, methanone, di-2-pyridinyl, 2,2'-dipyridyl ketone, 2-pyridine-2-carbonyl pyridine, pyridyl ketone
PubChem CID 88065
IUPAC Name dipyridin-2-ylmethanone
SMILES C1=CC=NC(=C1)C(=O)C2=CC=CC=N2

Specifications

Melting Point 53°C to 55°C
Color Colorless to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CO(C5H4N)
Quantity 5 g
Beilstein 24, IV, 591
Solubility Information Solubility in water: immiscible
Formula Weight 184.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Di-2-pyridyl ketone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 243-067-3

RUO – Research Use Only