missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-1-adamantylphosphine 95.0+%, TCI America™
$80.16 - $234.87
Chemical Identifiers
| CAS | 131211-27-3 |
|---|---|
| Molecular Formula | C20H31P |
| Molecular Weight (g/mol) | 302.44 |
| MDL Number | MFCD06658117 |
| InChI Key | RRRZOLBZYZWQBZ-UHFFFAOYSA-N |
| PubChem CID | 11077518 |
| IUPAC Name | bis(adamantan-1-yl)phosphane |
| SMILES | C1C2CC3CC1CC(C2)(C3)PC12CC3CC(CC(C3)C1)C2 |
Chemical Identifiers
| 131211-27-3 | |
| 302.44 | |
| RRRZOLBZYZWQBZ-UHFFFAOYSA-N | |
| bis(adamantan-1-yl)phosphane |
| C20H31P | |
| MFCD06658117 | |
| 11077518 | |
| C1C2CC3CC1CC(C2)(C3)PC12CC3CC(CC(C3)C1)C2 |
Specifications
| 131211-27-3 | |
| White | |
| MFCD06658117 | |
| RRRZOLBZYZWQBZ-UHFFFAOYSA-N | |
| bis(adamantan-1-yl)phosphane | |
| 11077518 | |
| ≥95.0% (GC) | |
| Di-1-adamantylphosphine |
| 178°C | |
| C20H31P | |
| 200 mg | |
| C1C2CC3CC1CC(C2)(C3)PC12CC3CC(CC(C3)C1)C2 | |
| 302.44 | |
| 302.44 | |
| Crystalline Powder |
Safety and Handling
TSCA : No