missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-1-adamantylphosphine 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 178°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 302.44 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Di-1-adamantylphosphine |
Safety and Handling
| TSCA | No |