missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ DGCR8 Knockout MEF Cells
Description
- Cells are supplied in 1.5mL quantities (about 1 x 106 cells/mL) in freezing media (80% MEF media, 20% FBS, 20% DMSO)
- Gene ID: 54487
- Gene symbol: DGCR8
Recommended Storage
Store in gas phase of liquid nitrogen
Warranty
6 Months
Specifications
Specifications
| Content And Storage | Store in gas phase of liquid nitrogen |
| For Use With (Application) | In vitro assay |
| Product Line | Novus |
| Product Type | DGCR8 Knockout MEF Cells |
| Quantity | 1.5 mL |
This product is for research use only and is not approved for use in humans or in clinical diagnosis