Learn More
Abcam Inc Carbo x yl-DCFDA (5(6)-Carbo x y-2',7'-dichlorofluorescein diacetate), Fluorogenic esterase substrate, 1G
Catalog No. 502705213
Encompass
MW 529.28 Da, Purity >85%. Amine-reactive, fluorogenic, non-selective esterase substrate. Hydrolyses to DFCA (ab145439) fluorophore intracellularly. Labels intact cells. 2,7-Dichlorofluorescein (ab146332) derivative and DCFDA N-succinimidyl ester (ab145286) precursor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705213 Supplier Abcam Inc Supplier No. AB1454151G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 529.28 Da, Purity >85%. Amine-reactive, fluorogenic, non-selective esterase substrate. Hydrolyses to DFCA (ab145439) fluorophore intracellularly. Labels intact cells. 2,7-Dichlorofluorescein (ab146332) derivative and DCFDA N-succinimidyl ester (ab145286) precursor.
The product is subject to the following: Abcam Restricted Use Statement