missing translation for 'onlineSavingsMsg'
Learn More

Dextrose U.S.P. GenAR Granular, Macron Fine Chemicals™

Catalog No. MK7730212
Change view
Click to view available options
Poly; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK7730212 Poly; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK7730212 Supplier Avantor Supplier No. 773006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 50-99-7
Packaging Poly
Molecular Formula C6H12O6
Quantity 2.5kg
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
SMILES C(C(C(C(C(C=O)O)O)O)O)O
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
Molecular Weight (g/mol) 180.156
PubChem CID 107526
ChEBI CHEBI:42758
Formula Weight 180.16
Grade USP
Chemical Name or Material Dextrose, Anhydrous
Show More Show Less