missing translation for 'onlineSavingsMsg'
Learn More

Dextrose Pharmaceutical Secondary Standard, 99.9%, MilliporeSigma™ Supelco™

Catalog No. 111014388
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014388 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014388 Supplier Merck Emd Millipore Supplier No. PHR10001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dextrose belongs to the class of sugars, and is a preferred source of carbohydrate in parenteral nutrition regimens. It is widely used in solutions to adjust tonicity, as a sweetening agent, wet granulation diluent/binder, direct-compression tablet diluent/binder, etc. It is considered only for short term clinical use, since it is deficient in electrolytes.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Food and Beverages and Pharmaceutical (small molecule)

Specifications

CAS 50-99-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula C6H12O6
MDL Number MFCD00063774
Melting Point 150°C to 152°C
Synonym D-(+)-Glucose; Dextrose
Type Certified Reference Material
Quantity 1 g
Formula Weight 180.16
Percent Purity 99.9%
Grade Certified Reference Material
Show More Show Less