missing translation for 'onlineSavingsMsg'
Learn More

Dextrose Impurity B, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022998
Change view
Click to view available options
:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111022998 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022998 Supplier Merck Emd Millipore Supplier No. PHR2085100MG
Only null left

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 499-40-1
Flash Point Not Applicable
Molecular Formula C12H22O11
Linear Formula C12H22O11
Quantity 100 mg
MDL Number MFCD00065373
Synonym Isomaltose; 6-O-alpha-D-Glucopyranosyl-D-glucose
Formula Weight 342.30
Grade Certified Reference Material