missing translation for 'onlineSavingsMsg'
Learn More

Dextran from Leuconostoc mesenteroides, For GPC, Mw 670,000, MilliporeSigma™ Supelco™

Catalog No. 111015817
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015817 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015817 Supplier Merck Emd Millipore Supplier No. 31425100MGF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manganese(II) sulfate monohydrate is a sulfate salt of manganese. Its toxic and carcinogenic effects have been evaluated in male and female F344/N rats and B6C3F1 mice.

Food and Beverages

Chemical Identifiers

MDL Number MFCD00130935

Specifications

Physical Form Neat
CAS 10034-96-5
Linear Formula (C6H10O5)n
Quantity 100 mg
Grade GPC