missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Dexterity™ Flame-Resistant Arc Flash Gloves with Neoprene Palm - 13 Gauge

Catalog No. 296365137
Change view
Click to view available options
Size:
10
11
6
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296365137 11
296365132 6
296365133 7
296365134 8
296365135 9
296365136 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 296365137 Supplier Superior Glove™ Supplier No. S13FRNE11

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specially engineered yarn blends Aramid Fibers with a variety of inherently flame resistant fibers for Arc Flash Level 2 protection. Neoprene coating on the palm enhances grip, and provides resistance to flames and oil.

  • Engineered yarn blends Aramid Fibers with a variety of strong and inherently flame-resistant fibers for arc flash protection
  • 13-gauge knit shell increases dexterity and hand movement
  • Neoprene palm coating enhances wet/dry grip, flame-resistant properties and provides resistance to oil
  • Arc flash testing placed these gloves at level 2, with an Arc Thermal Performance Value (ATPV) of 11 cal/cm2
  • Gloves are treated with Ultra-Fresh to achieve 99.9% antimicrobial status
  • Touchscreen compatible

Specifications

ANSI Abrasion Rating ANSI 4
Arc Flash Level 2
Coating Material Neoprene
Color Gray
Cut Resistance Rating ANSI A4
Material Aramid Fiber
Product Type Arc Flash Gloves
Size 11
For Use With (Application) Utilities, Petrochemical, Oil and Gas Industries
Glove Cuff Style Knit
Series S13FRNE
Yarn Gauge 13
Show More Show Less