missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Dexterity™ Flame-Resistant Arc Flash Gloves with Neoprene Palm - 13 Gauge

Catalog No. 296365135
Change view
Click to view available options
Size:
10
11
6
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296365135 9
296365132 6
296365133 7
296365134 8
296365136 10
296365137 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 296365135 Supplier Superior Glove™ Supplier No. S13FRNE9
Only null left

Specially engineered yarn blends Aramid Fibers with a variety of inherently flame resistant fibers for Arc Flash Level 2 protection. Neoprene coating on the palm enhances grip, and provides resistance to flames and oil.

  • Engineered yarn blends Aramid Fibers with a variety of strong and inherently flame-resistant fibers for arc flash protection
  • 13-gauge knit shell increases dexterity and hand movement
  • Neoprene palm coating enhances wet/dry grip, flame-resistant properties and provides resistance to oil
  • Arc flash testing placed these gloves at level 2, with an Arc Thermal Performance Value (ATPV) of 11 cal/cm2
  • Gloves are treated with Ultra-Fresh to achieve 99.9% antimicrobial status
  • Touchscreen compatible

Specifications

ANSI Abrasion Rating ANSI 4
Arc Flash Level 2
Coating Material Neoprene
Color Gray
Cut Resistance Rating ANSI A4
Material Aramid Fiber
Product Type Arc Flash Gloves
Size 9
For Use With (Application) Utilities, Petrochemical, Oil and Gas Industries
Glove Cuff Style Knit
Series S13FRNE
Yarn Gauge 13
Show More Show Less