missing translation for 'onlineSavingsMsg'
Learn More

Dexpanthenol, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18602073
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
45 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602073 45 kg Amber Glass Bottle
18602070 25 g Amber Glass Bottle
18602071 100 g Amber Glass Bottle
18602072 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602073 Supplier Spectrum Chemical Mfg Cor Supplier No. P183145KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1831, 81-13-0, C9H19NO4

Spectrum™ Chemical Dexpanthenol, USP acts on the gastrointestinal tract and increases lower intestinal motility. It is also applied topically to the skin to relieve itching and to promote healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 81-13-0
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00065006
InChI Key SNPLKNRPJHDVJA-ZETCQYMHSA-N
IUPAC Name (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)[C@@H](O)C(=O)NCCCO

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Refractive Index 1.495-1.502
Quantity 45 kg
Specific Rotation +29.0° to +31.5°
Percent Purity 98 to 102%
Grade USP
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Dexpanthenol