missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexpanthenol, USP, 98-102%, Spectrum™ Chemical
Description
Spectrum™ Chemical Dexpanthenol, USP acts on the gastrointestinal tract and increases lower intestinal motility. It is also applied topically to the skin to relieve itching and to promote healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.1% |
| Packaging | Amber Glass Bottle |
| Refractive Index | 1.495-1.502 |
| Quantity | 25 g |
| Specific Rotation | +29.0° to +31.5° |
| Percent Purity | 98 to 102% |
| Grade | USP |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Dexpanthenol |