missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexpanthenol, FCC, 98-102%, Spectrum™ Chemical
PA118, 81-13-0, C9H19NO4
Supplier: Spectrum Chemical Mfg Cor PA118100GM
Description
Spectrum™ Chemical Dexpanthenol, FCC is the provitamin of vitamin B5 and is used in nutritional supplements. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 81-13-0 | |
| 0.1% | |
| C9H19NO4 | |
| MFCD00065006 | |
| SNPLKNRPJHDVJA-ZETCQYMHSA-N | |
| CC(C)(CO)[C@@H](O)C(=O)NCCCO | |
| 205.25 | |
| FCC |
| 100% | |
| Amber Glass Bottle | |
| 1.495-1.502 | |
| 100 g | |
| +29.0° to +31.5° | |
| (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide | |
| 98 to 102% |
Chemical Identifiers
| 81-13-0 | |
| 205.25 | |
| SNPLKNRPJHDVJA-ZETCQYMHSA-N | |
| CC(C)(CO)[C@@H](O)C(=O)NCCCO |
| C9H19NO4 | |
| MFCD00065006 | |
| (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide |