missing translation for 'onlineSavingsMsg'
Learn More

Dexpanthenol, FCC, 98-102%, Spectrum™ Chemical

Catalog No. 18602066
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
45 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602066 25 g Amber Glass Bottle
18602067 100 g Amber Glass Bottle
18602068 1 kg Amber Glass Bottle
18602069 45 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602066 Supplier Spectrum Chemical Mfg Cor Supplier No. PA11825GM
Get Up to $500 in Chemicals with Purchase
View all promotions

PA118, 81-13-0, C9H19NO4

Spectrum™ Chemical Dexpanthenol, FCC is the provitamin of vitamin B5 and is used in nutritional supplements. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 81-13-0
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00065006
InChI Key SNPLKNRPJHDVJA-ZETCQYMHSA-N
IUPAC Name (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)[C@@H](O)C(=O)NCCCO

Specifications

CAS Max % 100%
Boiling Point 118°C to 120°C (244.4°F to 248°F) at 0.02 mmHg with decomposition
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Refractive Index 1.495-1.502
Quantity 25 g
Optical Rotation +29.0° to +31.5°
Percent Purity 98 to 102%
Grade FCC
Physical Form Liquid
Chemical Name or Material Dexpanthenol
Show More Show Less