Learn More
Thermo Scientific Chemicals Dexmedetomidine hydrochloride
$339.46 - $339.46
Chemical Identifiers
| CAS | 145108-58-3 |
|---|---|
| Molecular Formula | C13H17ClN2 |
| Molecular Weight (g/mol) | 236.74 |
| InChI Key | VPNGEIHDPSLNMU-MERQFXBCSA-N |
| IUPAC Name | hydrogen 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole chloride |
| SMILES | [H+].[Cl-].C[C@H](C1=CN=CN1)C1=CC=CC(C)=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465432500
|
Thermo Scientific Chemicals
465432500 |
250 mg |
Each for $339.46
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 145108-58-3 | |
| 236.74 | |
| hydrogen 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole chloride |
| C13H17ClN2 | |
| VPNGEIHDPSLNMU-MERQFXBCSA-N | |
| [H+].[Cl-].C[C@H](C1=CN=CN1)C1=CC=CC(C)=C1C |
Specifications
| 145108-58-3 | |
| C13H17ClN2 | |
| VPNGEIHDPSLNMU-MERQFXBCSA-N | |
| hydrogen 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole chloride | |
| 236.74 |
| 158.0°C | |
| 250 mg | |
| [H+].[Cl-].C[C@H](C1=CN=CN1)C1=CC=CC(C)=C1C | |
| 236.74 | |
| Dexmedetomidine hydrochloride |
Safety and Handling
GHS H Statement Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO – Research Use Only